missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein

A DNA sequence encoding the Influenza A virus (strain A/England/878/1969 H3N2) Influenza A H3N2 (strain A/England/878/1969) Hemagglutinin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7283-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018000

  • 2985.00 EUR / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein
Research Use Only
HA
Recombinant Protein
E. coli
QDLPGNDNST ATLCLGHHAV PNGTLVKTIT NDQIEVTNAT ELVQSSSTGK ICNNPHRILD GINCTLIDAL LGDPHCDVFQ DETWDLFVER SKAFSNCYPY DVPDYASLRS LVASSGTLEF ITEGFTWTGV TQNGGSNACK RGPDSGFFSR LNWLTKSGST YPVLNVTMPN NDNFDKLYIW GVHHPSTNQE QTSLYVQASG RVTVSTRRSQ QTIIPNIGSR PWVRGLSSRI SIYWTIVKPG DVLVINSNGN LIAPRGYFKM RTGKSSIMRS DAPIDTCISE CITPNGSIPN DKPFQNVNKI TYGACPKYVK QNTLKLATGM RNVPEKQT
Greater than 90% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Influenza
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers