missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/England/878/1969 H3N2) Influenza A H3N2 (strain A/England/878/1969) Hemagglutinin, was expressed in the hosts and tags indicated.
309.00 EUR - 2985.00 EUR
Specifications
Name | Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | HA |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097990
|
enQuireBio™
QP7283-EC-100UG |
100 μg |
865.00 EUR
100µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018000
|
enQuireBio™
QP7283-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008000
|
enQuireBio™
QP7283-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028000
|
enQuireBio™
QP7283-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038000
|
enQuireBio™
QP7283-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048000
|
enQuireBio™
QP7283-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Influenza A H3N2 (England / 878 / 1969) Hemagglutinin Protein | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
QDLPGNDNST ATLCLGHHAV PNGTLVKTIT NDQIEVTNAT ELVQSSSTGK ICNNPHRILD GINCTLIDAL LGDPHCDVFQ DETWDLFVER SKAFSNCYPY DVPDYASLRS LVASSGTLEF ITEGFTWTGV TQNGGSNACK RGPDSGFFSR LNWLTKSGST YPVLNVTMPN NDNFDKLYIW GVHHPSTNQE QTSLYVQASG RVTVSTRRSQ QTIIPNIGSR PWVRGLSSRI SIYWTIVKPG DVLVINSNGN LIAPRGYFKM RTGKSSIMRS DAPIDTCISE CITPNGSIPN DKPFQNVNKI TYGACPKYVK QNTLKLATGM RNVPEKQT | |
Greater than 90% as determined by SDS-PAGE. |