missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human WNT1-inducible-signaling pathway protein 1

A DNA sequence encoding the Homo sapiens (Human) WNT1-inducible-signaling pathway protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6886-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16086550

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001310298.1
WNT1-inducible-signaling pathway protein 1
Research Use Only
WISP2
Recombinant Protein
E. coli
QLCPTPCTCP WPPPRCPLGV PLVLDGCGCC RVCARRLGEP CDQLHVCDAS QGLVCQPGAG PGGRGALCLL AEDDSSCEVN GRLYREGETF QPHCSIRCRC EDGGFTCVPL CSEDVRLPSW DCPHPRRVEV LGKCCPEWVC GQGGGLGTQP LPAQGPQFSG LVSSLPPGVP CPEWSTAWGP CSTTCGLGMA TRVSNQNRFC RLETQRRLCL SRPCPPSRGR SPQNSAF
Greater than 80% as determined by SDS-PAGE.
8839
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers