missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WNT1-inducible-signaling pathway protein 1
A DNA sequence encoding the Homo sapiens (Human) WNT1-inducible-signaling pathway protein 2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001310298.1 |
---|---|
Gene ID (Entrez) | 8839 |
Name | WNT1-inducible-signaling pathway protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16076550
|
enQuireBio™
QP6886-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096550
|
enQuireBio™
QP6886-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16086550
|
enQuireBio™
QP6886-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006560
|
enQuireBio™
QP6886-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016560
|
enQuireBio™
QP6886-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026560
|
enQuireBio™
QP6886-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001310298.1 | |
WNT1-inducible-signaling pathway protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
8839 | |
Research Use Only | |
WISP2 | |
Recombinant Protein | |
E. coli | |
QLCPTPCTCP WPPPRCPLGV PLVLDGCGCC RVCARRLGEP CDQLHVCDAS QGLVCQPGAG PGGRGALCLL AEDDSSCEVN GRLYREGETF QPHCSIRCRC EDGGFTCVPL CSEDVRLPSW DCPHPRRVEV LGKCCPEWVC GQGGGLGTQP LPAQGPQFSG LVSSLPPGVP CPEWSTAWGP CSTTCGLGMA TRVSNQNRFC RLETQRRLCL SRPCPPSRGR SPQNSAF | |
Greater than 80% as determined by SDS-PAGE. |