missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WNT1-inducible-signaling pathway protein 1
A DNA sequence encoding the Homo sapiens (Human) WNT1-inducible-signaling pathway protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6886-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001310298.1 | |
WNT1-inducible-signaling pathway protein 1 | |
Research Use Only | |
WISP2 | |
Recombinant Protein | |
E. coli | |
QLCPTPCTCP WPPPRCPLGV PLVLDGCGCC RVCARRLGEP CDQLHVCDAS QGLVCQPGAG PGGRGALCLL AEDDSSCEVN GRLYREGETF QPHCSIRCRC EDGGFTCVPL CSEDVRLPSW DCPHPRRVEV LGKCCPEWVC GQGGGLGTQP LPAQGPQFSG LVSSLPPGVP CPEWSTAWGP CSTTCGLGMA TRVSNQNRFC RLETQRRLCL SRPCPPSRGR SPQNSAF | |
Greater than 80% as determined by SDS-PAGE. |
8839 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |