missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Wnt/RSPO2 Agonist Protein, CF

Product Code. 30241021
Click to view available options
Quantity:
1 mg
100 μg
50 μg
500 μg
Unit Size:
100µg
1mg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30241021

Brand: R&D Systems™ BTWRSP2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its ability to activate TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 6.00-72.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) 83-97 kDa & >190 kDa under reducing conditions.
Quantity 500 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Wnt/RSPO2 protein Proprietary
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CRISTIN2, HHRRD, Roof plate-specific spondin-2, R-spondin 2 homolog, R-spondin-2, TETAMS2, Wnt
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500μg/mL in PBS.
Form Lyophilized
Protein Wnt/RSPO2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.