missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 Q2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 Q2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7927-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138807.1 | |
Ubiquitin-conjugating enzyme E2 Q2 Protein | |
Research Use Only | |
UBE2Q2 | |
Recombinant Protein | |
E. coli | |
MSVSGLKAEL KFLASIFDKN HERFRIVSWK LDELHCQFLV PQQGSPHSLP PPLTLHCNIT ESYPSSSPIW FVDSEDPNLT SVLERLEDTK NNNLLRQQLK WLICELCSLY NLPKHLDVEM LDQPLPTGQN GTTEEVTSEE EEEEEEMAED IEDLDHYEMK EEEPISGKKS EDEGIEKENL AILEKIRKTQ RQDHLNGAVS GSVQASDRLM KELRDIYRSQ SYKTGIYSVE LINDSLYDWH VKLQKVDPDS PLHSDLQILK EKEGIEYILL NFSFKDNFPF DPPFVRVVLP VLSGGYVLGG GALCMELLTK QGWSSAYSIE SVIMQINATL VKGKARVQFG ANKNQYNLAR AQQSYNSIVQ IHEKNGWYTP PKEDG | |
Greater than 90% as determined by SDS-PAGE. |
92912 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |