missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ubiquitin-conjugating enzyme E2 Q2 Protein
A DNA sequence encoding the Homo sapiens (Human) Ubiquitin-conjugating enzyme E2 Q2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001138807.1 |
---|---|
Gene ID (Entrez) | 92912 |
Name | Ubiquitin-conjugating enzyme E2 Q2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031641
|
enQuireBio™
QP7927-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051641
|
enQuireBio™
QP7927-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041641
|
enQuireBio™
QP7927-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061641
|
enQuireBio™
QP7927-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071641
|
enQuireBio™
QP7927-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081641
|
enQuireBio™
QP7927-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001138807.1 | |
Ubiquitin-conjugating enzyme E2 Q2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
92912 | |
Research Use Only | |
UBE2Q2 | |
Recombinant Protein | |
E. coli | |
MSVSGLKAEL KFLASIFDKN HERFRIVSWK LDELHCQFLV PQQGSPHSLP PPLTLHCNIT ESYPSSSPIW FVDSEDPNLT SVLERLEDTK NNNLLRQQLK WLICELCSLY NLPKHLDVEM LDQPLPTGQN GTTEEVTSEE EEEEEEMAED IEDLDHYEMK EEEPISGKKS EDEGIEKENL AILEKIRKTQ RQDHLNGAVS GSVQASDRLM KELRDIYRSQ SYKTGIYSVE LINDSLYDWH VKLQKVDPDS PLHSDLQILK EKEGIEYILL NFSFKDNFPF DPPFVRVVLP VLSGGYVLGG GALCMELLTK QGWSSAYSIE SVIMQINATL VKGKARVQFG ANKNQYNLAR AQQSYNSIVQ IHEKNGWYTP PKEDG | |
Greater than 90% as determined by SDS-PAGE. |