missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMM17B Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM17B, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_001161419.1 |
---|---|
Gene ID (Entrez) | 10245 |
Name | TIMM17B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015890
|
enQuireBio™
QP6792-EC-100UG |
100 μg |
675.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035890
|
enQuireBio™
QP6792-EC-1MG |
1 mg |
2985.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025890
|
enQuireBio™
QP6792-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045890
|
enQuireBio™
QP6792-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055890
|
enQuireBio™
QP6792-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065890
|
enQuireBio™
QP6792-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001161419.1 | |
TIMM17B Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10245 | |
Research Use Only | |
TIMM17B | |
Recombinant Protein | |
E. coli | |
MEEYAREPCP WRIVDDCGGA FTMGVIGGGV FQAIKGFRNA PVGIRHRLRG SANAVRIRAP QIGGSFAVWG GLFSTIDCGL VRLRGKEDPW NSITSGALTG AVLAARSGPL AMVGSAMMGG ILLALIEGVG ILLTRYTAQQ FRNAPPFLED PSQLPPKDGT PAPGYPSYQQ YH | |
Greater than 80% as determined by SDS-PAGE. |