missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMM17B Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM17B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6792-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001161419.1 | |
TIMM17B Protein | |
Research Use Only | |
TIMM17B | |
Recombinant Protein | |
E. coli | |
MEEYAREPCP WRIVDDCGGA FTMGVIGGGV FQAIKGFRNA PVGIRHRLRG SANAVRIRAP QIGGSFAVWG GLFSTIDCGL VRLRGKEDPW NSITSGALTG AVLAARSGPL AMVGSAMMGG ILLALIEGVG ILLTRYTAQQ FRNAPPFLED PSQLPPKDGT PAPGYPSYQQ YH | |
Greater than 80% as determined by SDS-PAGE. |
10245 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |