missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIM3 Protein
A DNA sequence encoding the Homo sapiens (Human) TIM3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6145-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116171.3 | |
TIM3 Protein | |
Research Use Only | |
HAVCR2 | |
Recombinant Protein | |
Yeast | |
SEVEYRAEVG QNAYLPCFYT PAAPGNLVPV CWGKGACPVF ECGNVVLRTD ERDVNYWTSR YWLNGDFRKG DVSLTIENVT LADSGIYCCR IQIPGIMNDE KFNLKLVIKP AKVTPAPTRQ RDFTAAFPRM LTTRGHGPAE TQTLGSLPDI NLTQISTLAN ELRDSRLAND LRDSGATIRI G | |
Greater than 90% as determined by SDS-PAGE. |
84868 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |