missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIM3 Protein
A DNA sequence encoding the Homo sapiens (Human) TIM3, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_116171.3 |
---|---|
Gene ID (Entrez) | 84868 |
Name | TIM3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091140
|
enQuireBio™
QP6145-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031150
|
enQuireBio™
QP6145-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001150
|
enQuireBio™
QP6145-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021150
|
enQuireBio™
QP6145-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041150
|
enQuireBio™
QP6145-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011150
|
enQuireBio™
QP6145-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_116171.3 | |
TIM3 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
84868 | |
Research Use Only | |
HAVCR2 | |
Recombinant Protein | |
Yeast | |
SEVEYRAEVG QNAYLPCFYT PAAPGNLVPV CWGKGACPVF ECGNVVLRTD ERDVNYWTSR YWLNGDFRKG DVSLTIENVT LADSGIYCCR IQIPGIMNDE KFNLKLVIKP AKVTPAPTRQ RDFTAAFPRM LTTRGHGPAE TQTLGSLPDI NLTQISTLAN ELRDSRLAND LRDSGATIRI G | |
Greater than 90% as determined by SDS-PAGE. |