missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TALLA-1 / TSPAN7 Protein

A DNA sequence encoding the Homo sapiens (Human) TALLA-1 / TSPAN7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9449-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018341

  • 1585.00 EUR / 500µg
Estimated Shipment: 23-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004606.2
TALLA-1 / TSPAN7 Protein
Research Use Only
TSPAN7
Recombinant Protein
Yeast
RHEIKDTFLR TYTDAMQTYN GNDERSRAVD HVQRSLSCCG VQNYTNWSTS PYFLEHGIPP SCCMNETDCN PQDLHNLTVA ATKVNQKGCY DLVTSFMETN MGIIAGVAFG I
Greater than 90% as determined by SDS-PAGE.
7102
500 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers