missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syndecan-1 / SDC1 / CD138 Protein
A DNA sequence encoding the Homo sapiens (Human) Syndecan-1 / SDC1 / CD138, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6655-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001006947.1 | |
Syndecan-1 / SDC1 / CD138 Protein | |
Research Use Only | |
SDC1 | |
Recombinant Protein | |
E. coli | |
QIVATNLPPE DQDGSGDDSD NFSGSGAGAL QDITLSQQTP STWKDTQLLT AIPTSPEPTG LEATAASTST LPAGEGPKEG EAVVLPEVEP GLTAREQEAT PRPRETTQLP TTHLASTTTA TTAQEPATSH PHRDMQPGHH ETSTPAGPSQ ADLHTPHTED GGPSATERAA EDGASSQLPA AEGSGEQDFT FETSGENTAV VAVEPDRRNQ SPVDQGATGA SQGLLDRKEV LG | |
Greater than 90% as determined by SDS-PAGE. |
6382 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |