missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Syndecan-1 / SDC1 / CD138 Protein

A DNA sequence encoding the Homo sapiens (Human) Syndecan-1 / SDC1 / CD138, was expressed in the hosts and tags indicated.

58.50 EUR - 2090.00 EUR

Specifications

Accession Number NP_001006947.1
Gene ID (Entrez) 6382
Name Syndecan-1 / SDC1 / CD138 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16054850
View Documents Promotion Details
enQuireBio™
QP6655-EC-50UG
50 μg
58.50 EUR
50µg
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16004850
View Documents Promotion Details
enQuireBio™
QP6655-EC-100UG
100 μg
463.00 EUR
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16024850
View Documents Promotion Details
enQuireBio™
QP6655-EC-1MG
1 mg
2090.00 EUR
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16034850
View Documents Promotion Details
enQuireBio™
QP6655-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16044850
View Documents Promotion Details
enQuireBio™
QP6655-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16014850
View Documents Promotion Details
enQuireBio™
QP6655-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001006947.1
Syndecan-1 / SDC1 / CD138 Protein
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
6382
Research Use Only
SDC1
Recombinant Protein
E. coli
QIVATNLPPE DQDGSGDDSD NFSGSGAGAL QDITLSQQTP STWKDTQLLT AIPTSPEPTG LEATAASTST LPAGEGPKEG EAVVLPEVEP GLTAREQEAT PRPRETTQLP TTHLASTTTA TTAQEPATSH PHRDMQPGHH ETSTPAGPSQ ADLHTPHTED GGPSATERAA EDGASSQLPA AEGSGEQDFT FETSGENTAV VAVEPDRRNQ SPVDQGATGA SQGLLDRKEV LG
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications