missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Steroid 21-hydroxylase Protein

Product Code. p-7152545
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15968579

Brand: enQuireBio™ QP5907ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Steroid 21-hydroxylase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000491.4
Gene ID (Entrez) 1589
Name Steroid 21-hydroxylase Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CYP21A2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MLLLGLLLLP LLAGARLLWN WWKLRSLHLP PLAPGFLHLL QPDLPIYLLG LTQKFGPIYR LHLGLQDVVV LNSKRTIEEA MVKKWADFAG RPEPLTYKLV SKNYPDLSLG DYSLLWKAHK KLTRSALLLG IRDSMEPVVE QLTQEFCERM RAQPGTPVAI EEEFSLLTCS IICYLTFGDK IKDDNLMPAY YKCIQEVLKT WSHWSIQIVD VIPFLRFFPN PGLRRLKQAI EKRDHIVEMQ LRQHKESLVA GQWRDMMDYM LQGVAQPSME EGSGQLLEGH VHMAAVDLLI GGTETTANTL SWAVVFLLHH PEIQQRLQEE LDHELGPGAS SSRVPYKDRA RLPLLNATIA EVLRLRPVVP LALPHRTTRP SSISGYDIPE GTVIIPNLQG AHLDETVWER PHEFWPDRFL EPGKNSRALA FGCGARVCLG EPLARLELFV VLTRLLQAFT LLPSGDALPS LQPLPHCSVI LKMQPFQVRL QPRGMGAHSP GQNQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.