missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine palmitoyltransferase 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine palmitoyltransferase 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6726-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035410

  • 2090.00 EUR / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004854.1
Serine palmitoyltransferase 2 Protein
Research Use Only
SPTLC2
Recombinant Protein
E. coli
RDFLRYWRIE KCHHATEREE QKDFVSLYQD FENFYTRNLY MRIRDNWNRP ICSVPGARVD IMERQSHDYN WSFKYTGNII KGVINMGSYN YLGFARNTGS CQEAAAKVLE EYGAGVCSTR QEIGNLDKHE ELEELVARFL GVEAAMAYGM GFATNSMNIP ALVGKGCLIL SDELNHASLV LGARLSGATI RIFKHNNMQS LEKLLKDAIV YGQPRTRRPW KKILILVEGI YSMEGSIVRL PEVIALKKKY KAYLYLDEAH SIGALGPTGR GVVEYFGLDP EDVDVMMGTF TKSFGASGGY IGGKKELIDY LRTHSHSAVY ATSLSPPVVE QIITSMKCIM GQDGTSLGKE CVQQLAENTR YFRRRLKEMG FIIYGNEDSP VVPLMLYMPA KIGAFGREML KRNIGVVVVG FPATPIIESR ARFCLSAAHT KEILDTALKE IDEVGDLLQL KYSRHRLVPL LDRPFDETTY EETED
Greater than 90% as determined by SDS-PAGE.
9517
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers