missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine palmitoyltransferase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine palmitoyltransferase 2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_004854.1 |
---|---|
Gene ID (Entrez) | 9517 |
Name | Serine palmitoyltransferase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16015410
|
enQuireBio™
QP6726-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035410
|
enQuireBio™
QP6726-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025410
|
enQuireBio™
QP6726-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045410
|
enQuireBio™
QP6726-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055410
|
enQuireBio™
QP6726-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065410
|
enQuireBio™
QP6726-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004854.1 | |
Serine palmitoyltransferase 2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9517 | |
Research Use Only | |
SPTLC2 | |
Recombinant Protein | |
E. coli | |
RDFLRYWRIE KCHHATEREE QKDFVSLYQD FENFYTRNLY MRIRDNWNRP ICSVPGARVD IMERQSHDYN WSFKYTGNII KGVINMGSYN YLGFARNTGS CQEAAAKVLE EYGAGVCSTR QEIGNLDKHE ELEELVARFL GVEAAMAYGM GFATNSMNIP ALVGKGCLIL SDELNHASLV LGARLSGATI RIFKHNNMQS LEKLLKDAIV YGQPRTRRPW KKILILVEGI YSMEGSIVRL PEVIALKKKY KAYLYLDEAH SIGALGPTGR GVVEYFGLDP EDVDVMMGTF TKSFGASGGY IGGKKELIDY LRTHSHSAVY ATSLSPPVVE QIITSMKCIM GQDGTSLGKE CVQQLAENTR YFRRRLKEMG FIIYGNEDSP VVPLMLYMPA KIGAFGREML KRNIGVVVVG FPATPIIESR ARFCLSAAHT KEILDTALKE IDEVGDLLQL KYSRHRLVPL LDRPFDETTY EETED | |
Greater than 90% as determined by SDS-PAGE. |