missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine palmitoyltransferase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine palmitoyltransferase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6726-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004854.1 | |
Serine palmitoyltransferase 2 Protein | |
Research Use Only | |
SPTLC2 | |
Recombinant Protein | |
E. coli | |
RDFLRYWRIE KCHHATEREE QKDFVSLYQD FENFYTRNLY MRIRDNWNRP ICSVPGARVD IMERQSHDYN WSFKYTGNII KGVINMGSYN YLGFARNTGS CQEAAAKVLE EYGAGVCSTR QEIGNLDKHE ELEELVARFL GVEAAMAYGM GFATNSMNIP ALVGKGCLIL SDELNHASLV LGARLSGATI RIFKHNNMQS LEKLLKDAIV YGQPRTRRPW KKILILVEGI YSMEGSIVRL PEVIALKKKY KAYLYLDEAH SIGALGPTGR GVVEYFGLDP EDVDVMMGTF TKSFGASGGY IGGKKELIDY LRTHSHSAVY ATSLSPPVVE QIITSMKCIM GQDGTSLGKE CVQQLAENTR YFRRRLKEMG FIIYGNEDSP VVPLMLYMPA KIGAFGREML KRNIGVVVVG FPATPIIESR ARFCLSAAHT KEILDTALKE IDEVGDLLQL KYSRHRLVPL LDRPFDETTY EETED | |
Greater than 90% as determined by SDS-PAGE. |
9517 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |