missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SEPSECS Protein
A DNA sequence encoding the Homo sapiens (Human) SEPSECS, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8798-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_058651.3 | |
SEPSECS Protein | |
Research Use Only | |
SEPSECS | |
Recombinant Protein | |
E. coli | |
SFAAGERLVS PAYVRQGCEA RRSHEHLIRL LLEKGKCPEN GWDESTLELF LHELAIMDSN NFLGNCGVGE REGRVASALV ARRHYRFIHG IGRSGDISAV QPKAAGSSLL NKITNSLVLD IIKLAGVHTV ANCFVVPMAT GMSLTLCFLT LRHKRPKAKY IIWPRIDQKS CFKSMITAGF EPVVIENVLE GDELRTDLKA VEAKVQELGP DCILCIHSTT SCFAPRVPDR LEELAVICAN YDIPHIVNNA YGVQSSKCMH LIQQGARVGR IDAFVQSLDK NFMVPVGGAI IAGFNDSFIQ EISKMYPGRA SASPSLDVLI TLLSLGSNGY KKLLKERKEM FSYLSNQIKK LSEAYNERLL HTPHNPISLA MTLKTLDEHR DKAVTQLGSM LFTRQVSGAR VVPLGSMQTV SGYTFRGFMS HTNNYPCAYL NAASAIGMKM QDVDLFI | |
Greater than 90% as determined by SDS-PAGE. |
51091 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |