Learn More
enQuireBio™ Recombinant Human SEPSECS Protein
A DNA sequence encoding the Homo sapiens (Human) SEPSECS, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_058651.3 |
---|---|
Gene ID (Entrez) | 51091 |
Name | SEPSECS Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16086521
|
enQuireBio™
QP8798-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006531
|
enQuireBio™
QP8798-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16096521
|
enQuireBio™
QP8798-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16016531
|
enQuireBio™
QP8798-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026531
|
enQuireBio™
QP8798-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036531
|
enQuireBio™
QP8798-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_058651.3 | |
SEPSECS Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
51091 | |
Research Use Only | |
SEPSECS | |
Recombinant Protein | |
E. coli | |
SFAAGERLVS PAYVRQGCEA RRSHEHLIRL LLEKGKCPEN GWDESTLELF LHELAIMDSN NFLGNCGVGE REGRVASALV ARRHYRFIHG IGRSGDISAV QPKAAGSSLL NKITNSLVLD IIKLAGVHTV ANCFVVPMAT GMSLTLCFLT LRHKRPKAKY IIWPRIDQKS CFKSMITAGF EPVVIENVLE GDELRTDLKA VEAKVQELGP DCILCIHSTT SCFAPRVPDR LEELAVICAN YDIPHIVNNA YGVQSSKCMH LIQQGARVGR IDAFVQSLDK NFMVPVGGAI IAGFNDSFIQ EISKMYPGRA SASPSLDVLI TLLSLGSNGY KKLLKERKEM FSYLSNQIKK LSEAYNERLL HTPHNPISLA MTLKTLDEHR DKAVTQLGSM LFTRQVSGAR VVPLGSMQTV SGYTFRGFMS HTNNYPCAYL NAASAIGMKM QDVDLFI | |
Greater than 90% as determined by SDS-PAGE. |