missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human SCF Protein
Click to view available options
Quantity:
1 mg
10 μg
200 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
1mg
200µg
50µg
Description
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 1-5 ng/mL.
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 4254 |
| Molecular Weight (g/mol) | M.W. (theroretical): 19 kDa |
| Name | SCF/c-kit Ligand |
| Quantity | 1 mg |
| Source | E. coli-derived human SCF/c-kit Ligand protein Glu26-Ala189,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction