Learn More
enQuireBio™ Recombinant Human RPA1 Protein
A DNA sequence encoding the Homo sapiens (Human) RPA1, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_002936.1 |
---|---|
Gene ID (Entrez) | 6117 |
Name | RPA1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024590
|
enQuireBio™
QP6618-EC-100UG |
100 μg |
525.00 EUR
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044590
|
enQuireBio™
QP6618-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034590
|
enQuireBio™
QP6618-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054590
|
enQuireBio™
QP6618-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064590
|
enQuireBio™
QP6618-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074590
|
enQuireBio™
QP6618-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002936.1 | |
RPA1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6117 | |
Research Use Only | |
RPA1 | |
Recombinant Protein | |
E. coli | |
VGQLSEGAIA AIMQKGDTNI KPILQVINIR PITTGNSPPR YRLLMSDGLN TLSSFMLATQ LNPLVEEEQL SSNCVCQIHR FIVNTLKDGR RVVILMELEV LKSAEAVGVK IGNPVPYNEG LGQPQVAPPA PAASPAASSR PQPQNGSSGM GSTVSKAYGA SKTFGKAAGP SLSHTSGGTQ SKVVPIASLT PYQSKWTICA RVTNKSQIRT WSNSRGEGKL FSLELVDESG EIRATAFNEQ VDKFFPLIEV NKVYYFSKGT LKIANKQFTA VKNDYEMTFN NETSVMPCED DHHLPTVQFD FTGIDDLENK SKDSLVDIIG ICKSYEDATK ITVRSNNREV AKRNIYLMDT SGKVVTATLW GEDADKFDGS RQPVLAIKGA RVSDFGGRSL SVLSSSTIIA NPDIPEAYKL RGWFDAEGQA LDGVSISDLK SGGVGGSNTN WKTLYEVKSE NLGQGDKPDY FSSVATVVYL RKENCMYQAC PTQDCNKKVI DQQNGLYRCE KCDTEFPNFK YRMILSVNIA DFQENQWVTC FQESAEAILG QNAAYLGELK DKNEQAFEEV FQNANFRSFI FRVRVKVETY NDESRIKATV MDVKPVDYRE YGRRLVMSIR RSALM | |
Greater than 90% as determined by SDS-PAGE. |