missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Relaxin-2 (B-29/A-24) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Sodium Acetate.
Lyophilized from a 0.2μm filtered solution in sodium acetate with BSA as a carrier protein.
Unit Size:
25µg
Description
The Recombinant Human Relaxin-2 (B-29/A-24) Protein is derived from E. coli. The Recombinant Human Relaxin-2 (B-29/A-24) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Sodium Acetate. |
| Gene ID (Entrez) | 6019 |
| Molecular Weight (g/mol) | 2.7 kDa (A chain); 3.3 kDa (B chain) |
| Name | Relaxin-2 (B-29/A-24) |
| Quantity | 25 μg |
| Source | E. coli-derived human Relaxin-2 protein Asp25-Ser53 (B chain) & Gln162-Cys185 (A chain) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction