missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein Wnt-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6888-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016570

  • 2090.00 EUR / 1mg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_110388.2
Protein Wnt-4 Protein
Research Use Only
WNT4
Recombinant Protein
E. coli
SNWLYLAKLS SVGSISEEET CEKLKGLIQR QVQMCKRNLE VMDSVRRGAQ LAIEECQYQF RNRRWNCSTL DSLPVFGKVV TQGTREAAFV YAISSAGVAF AVTRACSSGE LEKCGCDRTV HGVSPQGFQW SGCSDNIAYG VAFSQSFVDV RERSKGASSS RALMNLHNNE AGRKAILTHM RVECKCHGVS GSCEVKTCWR AVPPFRQVGH ALKEKFDGAT EVEPRRVGSS RALVPRNAQF KPHTDEDLVY LEPSPDFCEQ DMRSGVLGTR GRTCNKTSKA IDGCELLCCG RGFHTAQVEL AERCSCKFHW CCFVKCRQCQ RLVELHTCR
Greater than 90% as determined by SDS-PAGE.
54361
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers