missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Wnt-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-4, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_110388.2 |
---|---|
Gene ID (Entrez) | 54361 |
Name | Protein Wnt-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16096560
|
enQuireBio™
QP6888-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16016570
|
enQuireBio™
QP6888-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16006570
|
enQuireBio™
QP6888-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16026570
|
enQuireBio™
QP6888-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036570
|
enQuireBio™
QP6888-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046570
|
enQuireBio™
QP6888-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_110388.2 | |
Protein Wnt-4 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
54361 | |
Research Use Only | |
WNT4 | |
Recombinant Protein | |
E. coli | |
SNWLYLAKLS SVGSISEEET CEKLKGLIQR QVQMCKRNLE VMDSVRRGAQ LAIEECQYQF RNRRWNCSTL DSLPVFGKVV TQGTREAAFV YAISSAGVAF AVTRACSSGE LEKCGCDRTV HGVSPQGFQW SGCSDNIAYG VAFSQSFVDV RERSKGASSS RALMNLHNNE AGRKAILTHM RVECKCHGVS GSCEVKTCWR AVPPFRQVGH ALKEKFDGAT EVEPRRVGSS RALVPRNAQF KPHTDEDLVY LEPSPDFCEQ DMRSGVLGTR GRTCNKTSKA IDGCELLCCG RGFHTAQVEL AERCSCKFHW CCFVKCRQCQ RLVELHTCR | |
Greater than 90% as determined by SDS-PAGE. |