missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein SSX4 Protein

Product Code. 16045460
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16045460 50 μg 50µg
16005460 10 μg 10µg
16095450 100 μg 100µg
16025460 200 μg 200µg
16035460 500 μg 500µg
16015460 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16045460 Supplier enQuireBio™ Supplier No. QP6733ec50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Protein SSX4, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001030004.1
Gene ID (Entrez) 548313
Name Protein SSX4 Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol SSX4
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MNGDDAFARR PRDDAQISEK LRKAFDDIAK YFSKKEWEKM KSSEKIVYVY MKLNYEVMTK LGFKVTLPPF MRSKRAADFH GNDFGNDRNH RNQVERPQMT FGSLQRIFPK IMPKKPAEEE NGLKEVPEAS GPQNDGKQLC PPGNPSTLEK INKTSGPKRG KHAWTHRLRE RKQLVVYEEI SDPEEDDE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.