missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nanos homolog 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Nanos homolog 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6402-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023060

  • 2095.00 EUR / 1mg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001025032.1
Nanos homolog 2 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
339345
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NANOS2
Recombinant Protein
E. coli
MQLPPFDMWK DYFNLSQVVW ALIASRGQRL ETQEIEEPSP GPPLGQDQGL GAPGANGGLG TLCNFCKHNG ESRHVYSSHQ LKTPDGVVVC PILRHYVCPV CGATGDQAHT LKYCPLNGGQ QSLYRRSGRN SAGRRVKR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers