missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nanos homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Nanos homolog 2, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001025032.1 |
---|---|
Gene ID (Entrez) | 339345 |
Name | Nanos homolog 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003060
|
enQuireBio™
QP6402-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023060
|
enQuireBio™
QP6402-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013060
|
enQuireBio™
QP6402-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033060
|
enQuireBio™
QP6402-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043060
|
enQuireBio™
QP6402-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053060
|
enQuireBio™
QP6402-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001025032.1 | |
Nanos homolog 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
339345 | |
Research Use Only | |
NANOS2 | |
Recombinant Protein | |
E. coli | |
MQLPPFDMWK DYFNLSQVVW ALIASRGQRL ETQEIEEPSP GPPLGQDQGL GAPGANGGLG TLCNFCKHNG ESRHVYSSHQ LKTPDGVVVC PILRHYVCPV CGATGDQAHT LKYCPLNGGQ QSLYRRSGRN SAGRRVKR | |
Greater than 90% as determined by SDS-PAGE. |