missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nanos homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Nanos homolog 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6402-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001025032.1 | |
Nanos homolog 2 Protein | |
Research Use Only | |
NANOS2 | |
Recombinant Protein | |
E. coli | |
MQLPPFDMWK DYFNLSQVVW ALIASRGQRL ETQEIEEPSP GPPLGQDQGL GAPGANGGLG TLCNFCKHNG ESRHVYSSHQ LKTPDGVVVC PILRHYVCPV CGATGDQAHT LKYCPLNGGQ QSLYRRSGRN SAGRRVKR | |
Greater than 90% as determined by SDS-PAGE. |
339345 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |