missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MUSK Kinase Protein
A DNA sequence encoding the Homo sapiens (Human) MUSK Kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6395-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001159752.1 | |
MUSK Kinase Protein | |
Research Use Only | |
MUSK | |
Recombinant Protein | |
E. coli | |
LPKAPVITTP LETVDALVEE VATFMCAVES YPQPEISWTR NKILIKLFDT RYSIRENGQL LTILSVEDSD DGIYCCTANN GVGGAVESCG ALQVKMKPKI TRPPINVKII EGLKAVLPCT TMGNPKPSVS WIKGDSPLRE NSRIAVLESG SLRIHNVQKE DAGQYRCVAK NSLGTAYSKV VKLEVEVFAR ILRAPESHNV TFGSFVTLHC TATGIPVPTI TWIENGNAVS SGSIQESVKD RVIDSRLQLF ITKPGLYTCI ATNKHGEKFS TAKAAATISI AEWSKPQKDN KGYCAQYRGE VCNAVLAKDA LVFLNTSYAD PEEAQELLVH TAWNELKVVS PVCRPAAEAL LCNHIFQECS PGVVPTPIPI CREYCLAVKE LFCAKEWLVM EEKTHRGLYR SEMHLLSVPE CSKLPSMHWD PTACARLPHL DYNKENLKTF PPMTSSKPSV DIPNLPSSSS SSFSVSPTYS MT | |
Greater than 90% as determined by SDS-PAGE. |
4593 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |