missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MUSK Kinase Protein
A DNA sequence encoding the Homo sapiens (Human) MUSK Kinase, was expressed in the hosts and tags indicated.
160.00 EUR - 2090.00 EUR
Specifications
Accession Number | NP_001159752.1 |
---|---|
Gene ID (Entrez) | 4593 |
Name | MUSK Kinase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16023010
|
enQuireBio™
QP6395-EC-100UG |
100 μg |
463.00 EUR
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043010
|
enQuireBio™
QP6395-EC-1MG |
1 mg |
2090.00 EUR
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033010
|
enQuireBio™
QP6395-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053010
|
enQuireBio™
QP6395-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063010
|
enQuireBio™
QP6395-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073010
|
enQuireBio™
QP6395-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001159752.1 | |
MUSK Kinase Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4593 | |
Research Use Only | |
MUSK | |
Recombinant Protein | |
E. coli | |
LPKAPVITTP LETVDALVEE VATFMCAVES YPQPEISWTR NKILIKLFDT RYSIRENGQL LTILSVEDSD DGIYCCTANN GVGGAVESCG ALQVKMKPKI TRPPINVKII EGLKAVLPCT TMGNPKPSVS WIKGDSPLRE NSRIAVLESG SLRIHNVQKE DAGQYRCVAK NSLGTAYSKV VKLEVEVFAR ILRAPESHNV TFGSFVTLHC TATGIPVPTI TWIENGNAVS SGSIQESVKD RVIDSRLQLF ITKPGLYTCI ATNKHGEKFS TAKAAATISI AEWSKPQKDN KGYCAQYRGE VCNAVLAKDA LVFLNTSYAD PEEAQELLVH TAWNELKVVS PVCRPAAEAL LCNHIFQECS PGVVPTPIPI CREYCLAVKE LFCAKEWLVM EEKTHRGLYR SEMHLLSVPE CSKLPSMHWD PTACARLPHL DYNKENLKTF PPMTSSKPSV DIPNLPSSSS SSFSVSPTYS MT | |
Greater than 90% as determined by SDS-PAGE. |