missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Muscarinic acetylcholine receptor M3 Protein
A DNA sequence encoding the Homo sapiens (Human) Muscarinic acetylcholine receptor M3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9067-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000731.1 | |
Muscarinic acetylcholine receptor M3 Protein | |
Research Use Only | |
CHRM3 | |
Recombinant Protein | |
Yeast | |
RIYKETEKRT KELAGLQASG TEAETENFVH PTGSSRSCSS YELQQQSMKR SNRRKYGRCH FWFTTKSWKP SSEQMDQDHS SSDSWNNNDA AASLENSASS DEEDIGSETR AIYSIVLKLP GHSTILNSTK LPSSDNLQVP EEELGMVDLE RKADKLQAQK SVDDGGSFPK SFSKLPIQLE SAVDTAKTSD VNSSVGKSTA TLPLSFKEAT LAKRFALKTR SQITKRKRMS LVKEKKAAQT | |
Greater than 90% as determined by SDS-PAGE. |
1131 | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |