missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Muscarinic acetylcholine receptor M3 Protein
A DNA sequence encoding the Homo sapiens (Human) Muscarinic acetylcholine receptor M3, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_000731.1 |
---|---|
Gene ID (Entrez) | 1131 |
Name | Muscarinic acetylcholine receptor M3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16077551
|
enQuireBio™
QP9067-YE-100UG |
100 μg |
640.00 EUR
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017561
|
enQuireBio™
QP9067-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087551
|
enQuireBio™
QP9067-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007561
|
enQuireBio™
QP9067-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027561
|
enQuireBio™
QP9067-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097551
|
enQuireBio™
QP9067-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000731.1 | |
Muscarinic acetylcholine receptor M3 Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1131 | |
Research Use Only | |
CHRM3 | |
Recombinant Protein | |
Yeast | |
RIYKETEKRT KELAGLQASG TEAETENFVH PTGSSRSCSS YELQQQSMKR SNRRKYGRCH FWFTTKSWKP SSEQMDQDHS SSDSWNNNDA AASLENSASS DEEDIGSETR AIYSIVLKLP GHSTILNSTK LPSSDNLQVP EEELGMVDLE RKADKLQAQK SVDDGGSFPK SFSKLPIQLE SAVDTAKTSD VNSSVGKSTA TLPLSFKEAT LAKRFALKTR SQITKRKRMS LVKEKKAAQT | |
Greater than 90% as determined by SDS-PAGE. |