missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human mRNA-decapping enzyme 1B Protein

A DNA sequence encoding the Homo sapiens(Human) mRNA-decapping enzyme 1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9835-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018831

  • 810.00 EUR / 100µg
Estimated Shipment: 03-02-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_689853.3
mRNA-decapping enzyme 1B Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
196513
Greater than 85% as determined by SDS-PAGE.
Research Use Only
DCP1B
Recombinant Protein
Yeast
AAVAAGGLVG KGRDISLAAL QRHDPYINRI VDVASQVALY TFGHRANEWE KTDVEGTLFV YTRSASPKHG FTIMNRLSME NRTEPITKDL DFQLQDPFLL YRNARLSIYG IWFYDKEECQ RIAELMKNLT QYEQLKAHQG TGAGISPVIL NSGEGKEVDI LRMLIKAKDE YTKCKTCSEP KKITSSSAIY DNPNLIKPIP VKPSENQQQR IPQPNQTLDP EPQHLSLTAL FGKQDKATCQ ETVEPPQTLH QQQQQQQQQQ EKLPIRQGVV RSLSYEEPRR HSPPIEKQLC PAIQKLMVRS ADLHPLSELP ENRPCENGST HSAGEFFTGP VQPGSPHNIG TSRGVQNASR TQNLFEKLQS TPGAANKCDP STPAPASSAA LNRSRAPTSV TPVAPGKGLA QPPQAYFNGS LPPQTVGHQA HGREQSTLPR QTLPISGSQT GSSGVISPQE LLKKLQIVQQ EQQLHASNRP ALAAKFPVLA QSSGTGKPLE SWINKTPNTE QQTPLFQVIS PQRIPATAAP SLLMSPMVFA QPTSVPPKER ESGLLPVGGQ EPPAAATSLL LPIQSPEPSV ITSSPLTKLQ LQEALLYLIQ NDDNFLNIIY EAYLFSMTQA AMKKTM
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers