missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human mRNA-decapping enzyme 1B Protein
A DNA sequence encoding the Homo sapiens(Human) mRNA-decapping enzyme 1B, was expressed in the hosts and tags indicated.
287.00 EUR - 3175.00 EUR
Specifications
Accession Number | NP_689853.3 |
---|---|
Gene ID (Entrez) | 196513 |
Name | mRNA-decapping enzyme 1B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16018831
|
enQuireBio™
QP9835-YE-100UG |
100 μg |
810.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16058831
|
enQuireBio™
QP9835-YE-500UG |
500 μg |
2325.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16028831
|
enQuireBio™
QP9835-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048831
|
enQuireBio™
QP9835-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068831
|
enQuireBio™
QP9835-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038831
|
enQuireBio™
QP9835-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_689853.3 | |
mRNA-decapping enzyme 1B Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
196513 | |
Research Use Only | |
DCP1B | |
Recombinant Protein | |
Yeast | |
AAVAAGGLVG KGRDISLAAL QRHDPYINRI VDVASQVALY TFGHRANEWE KTDVEGTLFV YTRSASPKHG FTIMNRLSME NRTEPITKDL DFQLQDPFLL YRNARLSIYG IWFYDKEECQ RIAELMKNLT QYEQLKAHQG TGAGISPVIL NSGEGKEVDI LRMLIKAKDE YTKCKTCSEP KKITSSSAIY DNPNLIKPIP VKPSENQQQR IPQPNQTLDP EPQHLSLTAL FGKQDKATCQ ETVEPPQTLH QQQQQQQQQQ EKLPIRQGVV RSLSYEEPRR HSPPIEKQLC PAIQKLMVRS ADLHPLSELP ENRPCENGST HSAGEFFTGP VQPGSPHNIG TSRGVQNASR TQNLFEKLQS TPGAANKCDP STPAPASSAA LNRSRAPTSV TPVAPGKGLA QPPQAYFNGS LPPQTVGHQA HGREQSTLPR QTLPISGSQT GSSGVISPQE LLKKLQIVQQ EQQLHASNRP ALAAKFPVLA QSSGTGKPLE SWINKTPNTE QQTPLFQVIS PQRIPATAAP SLLMSPMVFA QPTSVPPKER ESGLLPVGGQ EPPAAATSLL LPIQSPEPSV ITSSPLTKLQ LQEALLYLIQ NDDNFLNIIY EAYLFSMTQA AMKKTM | |
Greater than 85% as determined by SDS-PAGE. |