missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP-9 / CLG4B Protein
A DNA sequence encoding the Homo sapiens (Human) MMP-9 / CLG4B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9281-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004985.2 | |
MMP-9 / CLG4B Protein | |
Research Use Only | |
MMP9 | |
Recombinant Protein | |
Yeast | |
FQTFEGDLKW HHHNITYWIQ NYSEDLPRAV IDDAFARAFA LWSAVTPLTF TRVYSRDADI VIQFGVAEHG DGYPFDGKDG LLAHAFPPGP GIQGDAHFDD DELWSLGKGV VVPTRFGNAD GAACHFPFIF EGRSYSACTT DGRSDGLPWC STTANYDTDD RFGFCPSERL YTQDGNADGK PCQFPFIFQG QSYSACTTDG RSDGYRWCAT TANYDRDKLF GFCPTRADST VMGGNSAGEL CVFPFTFLGK EYSTCTSEGR GDGRLWCATT SNFDSDKKWG FCPDQGYSLF LVAAHEFGHA LGLDHSSVPE ALMYPMYRFT EGPPLHKDDV NGIRHLYGPR PEPEPRPPTT TTPQPTAPPT VCPTGPPTVH PSERPTAGPT GPPSAGPTGP PTAGPSTATT VPLSPVDDAC NVNIFDAIAE IGNQLYLFKD GKYWRFSEGR GSRPQGPFLI ADKWPALPRK LDSVFEERLS KKLFFFSGRQ VWVYTGASVL GPRRLDKLGL GADVAQVTGA LRSGRGKMLL FSGRRLWRFD VKAQMVDPRS ASEVDRMFPG VPLDTHDVFQ YREKAYFCQD RFYWRVSSRS ELNQVDQVGY VTYDILQCPE D | |
Greater than 90% as determined by SDS-PAGE. |
4318 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |