missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMP-9 / CLG4B Protein

A DNA sequence encoding the Homo sapiens (Human) MMP-9 / CLG4B, was expressed in the hosts and tags indicated.

202.00 EUR - 2155.00 EUR

Specifications

Accession Number NP_004985.2
Gene ID (Entrez) 4318
Name MMP-9 / CLG4B Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16018001
View Documents Promotion Details
enQuireBio™
QP9281-YE-100UG
100 μg
565.00 EUR
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16058001
View Documents Promotion Details
enQuireBio™
QP9281-YE-500UG
500 μg
1525.00 EUR
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16028001
View Documents Promotion Details
enQuireBio™
QP9281-YE-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16048001
View Documents Promotion Details
enQuireBio™
QP9281-YE-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16068001
View Documents Promotion Details
enQuireBio™
QP9281-YE-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16038001
View Documents Promotion Details
enQuireBio™
QP9281-YE-1MG
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_004985.2
MMP-9 / CLG4B Protein
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
4318
Research Use Only
MMP9
Recombinant Protein
Yeast
FQTFEGDLKW HHHNITYWIQ NYSEDLPRAV IDDAFARAFA LWSAVTPLTF TRVYSRDADI VIQFGVAEHG DGYPFDGKDG LLAHAFPPGP GIQGDAHFDD DELWSLGKGV VVPTRFGNAD GAACHFPFIF EGRSYSACTT DGRSDGLPWC STTANYDTDD RFGFCPSERL YTQDGNADGK PCQFPFIFQG QSYSACTTDG RSDGYRWCAT TANYDRDKLF GFCPTRADST VMGGNSAGEL CVFPFTFLGK EYSTCTSEGR GDGRLWCATT SNFDSDKKWG FCPDQGYSLF LVAAHEFGHA LGLDHSSVPE ALMYPMYRFT EGPPLHKDDV NGIRHLYGPR PEPEPRPPTT TTPQPTAPPT VCPTGPPTVH PSERPTAGPT GPPSAGPTGP PTAGPSTATT VPLSPVDDAC NVNIFDAIAE IGNQLYLFKD GKYWRFSEGR GSRPQGPFLI ADKWPALPRK LDSVFEERLS KKLFFFSGRQ VWVYTGASVL GPRRLDKLGL GADVAQVTGA LRSGRGKMLL FSGRRLWRFD VKAQMVDPRS ASEVDRMFPG VPLDTHDVFQ YREKAYFCQD RFYWRVSSRS ELNQVDQVGY VTYDILQCPE D
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications