missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Macrophage mannose receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Macrophage mannose receptor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6378-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002429.1
Macrophage mannose receptor 1 Protein
Research Use Only
MRC1
Recombinant Protein
Yeast
RTSLCFKLYA KGKHEKKTWF ESRDFCRALG GDLASINNKE EQQTIWRLIT ASGSYHKLFW LGLTYGSPSE GFTWSDGSPV SYENWAYGEP NNYQNVEYCG ELKGDPTMSW NDINCEHLNN WICQIQKGQT PKPEPTPAPQ DNPPVTEDGW VIYKDYQYYF SKEKETMDNA RAFCKRNFGD LVSIQSESEK KFLWKYVNRN DAQSAYFIGL LISLDKKFAW MDGSKVDYVS WATGEPNFAN EDENCVTMYS NSGFWNDINC GYPNAFICQR HNSSINATTV MPTMPSVPSG CKEGWNFYSN KCFKIFGFME EERKNWQEAR KACIGFGGNL VSIQNEKEQA FLTYHMKDST FSAWTGLNDV NSEHTFLWTD GRGVHYTNWG KGYPGGRRSS LSYEDADCVV IIGGASNEAG KWMDDTCDSK RGYICQTRSD PSLTNPPATI QTDGFVKYGK SSYSLMRQKF QWHEAETYCK LHNSLIASIL DPYSNAFAWL QMETSNERVW IALNSNLTDN QYTWTDKWRV RYTNWAADEP KLKSACVYLD LDGYWKTAHC NESFYFLCK
Greater than 90% as determined by SDS-PAGE.
4360
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers