missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Macrophage mannose receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Macrophage mannose receptor 1, was expressed in the hosts and tags indicated.
202.00 EUR - 2425.00 EUR
Specifications
Accession Number | NP_002429.1 |
---|---|
Gene ID (Entrez) | 4360 |
Name | Macrophage mannose receptor 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002880
|
enQuireBio™
QP6378-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022880
|
enQuireBio™
QP6378-YE-1MG |
1 mg |
2425.00 EUR
1mg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012880
|
enQuireBio™
QP6378-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032880
|
enQuireBio™
QP6378-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042880
|
enQuireBio™
QP6378-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052880
|
enQuireBio™
QP6378-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002429.1 | |
Macrophage mannose receptor 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4360 | |
Research Use Only | |
MRC1 | |
Recombinant Protein | |
Yeast | |
RTSLCFKLYA KGKHEKKTWF ESRDFCRALG GDLASINNKE EQQTIWRLIT ASGSYHKLFW LGLTYGSPSE GFTWSDGSPV SYENWAYGEP NNYQNVEYCG ELKGDPTMSW NDINCEHLNN WICQIQKGQT PKPEPTPAPQ DNPPVTEDGW VIYKDYQYYF SKEKETMDNA RAFCKRNFGD LVSIQSESEK KFLWKYVNRN DAQSAYFIGL LISLDKKFAW MDGSKVDYVS WATGEPNFAN EDENCVTMYS NSGFWNDINC GYPNAFICQR HNSSINATTV MPTMPSVPSG CKEGWNFYSN KCFKIFGFME EERKNWQEAR KACIGFGGNL VSIQNEKEQA FLTYHMKDST FSAWTGLNDV NSEHTFLWTD GRGVHYTNWG KGYPGGRRSS LSYEDADCVV IIGGASNEAG KWMDDTCDSK RGYICQTRSD PSLTNPPATI QTDGFVKYGK SSYSLMRQKF QWHEAETYCK LHNSLIASIL DPYSNAFAWL QMETSNERVW IALNSNLTDN QYTWTDKWRV RYTNWAADEP KLKSACVYLD LDGYWKTAHC NESFYFLCK | |
Greater than 90% as determined by SDS-PAGE. |