missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human m7GpppN-mRNA hydrolase Protein

Product Code. 16090921
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16090921 100 μg 100µg
16000931 10 μg 10µg
16040931 50 μg 50µg
16020931 200 μg 200µg
16030931 500 μg 500µg
16010931 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16090921 Supplier enQuireBio™ Supplier No. QP7809ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) m7GpppN-mRNA hydrolase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001229306.1
Gene ID (Entrez) 167227
Name m7GpppN-mRNA hydrolase Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol DCP2
Protein Length Full Length of Isoform 2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence METKRVEIPG SVLDDFCSRF ILHIPSEERD NAIRVCFQIE LAHWFYLDFY MQNTPGLPQC GIRDFAKAVF SHCPFLLPQG EDVEKVLDEW KEYKMGVPTY GAIILDETLE NVLLVQGYLA KSGWGFPKGK VNKEEAPHDC AAREVFEETG FDIKDYICKD DYIELRINDQ LARLYIIPGI PKDTKFNPKT RREIRNIEWF SIEKLPCHRN DMTPKSKLGL APNKFFMAIP FIRPLRDWLS RRFGDSSDSD NGFSSTGSTP AKPTVEKLSR TKFRHSQQLF PDGSPGDQWV KHRQPLQQKP YNNHSEMSDL LKGKKCEKKL HPRKLQDNFE TDAVYDLPSS SEDQLLEHAE GQPVACNGHC KFPFSSRAFL SFKFDHNAIM KILDL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.