missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRP2 Protein
A DNA sequence encoding the Homo sapiens (Human) LRP2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9256-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004516.2 | |
LRP2 Protein | |
Research Use Only | |
LRP2 | |
Recombinant Protein | |
Yeast | |
NCTASQFKCA SGDKCIGVTN RCDGVFDCSD NSDEAGCPTR PPGMCHSDEF QCQEDGICIP NFWECDGHPD CLYGSDEHNA CVPKTCPSSY FHCDNGNCIH RAWLCDRDND CGDMSDEKDC PTQPFRCPSW QWQCLGHNIC VNLSVVCDGI FDCPNGTDES PLCNGNSCSD FNGGCTHECV QEPFGAKCLC PLGFLLANDS KTCE | |
Greater than 90% as determined by SDS-PAGE. |
4036 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |