missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRP2 Protein
A DNA sequence encoding the Homo sapiens (Human) LRP2, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_004516.2 |
---|---|
Gene ID (Entrez) | 4036 |
Name | LRP2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16067921
|
enQuireBio™
QP9256-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007931
|
enQuireBio™
QP9256-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077921
|
enQuireBio™
QP9256-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16097921
|
enQuireBio™
QP9256-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017931
|
enQuireBio™
QP9256-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087921
|
enQuireBio™
QP9256-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004516.2 | |
LRP2 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4036 | |
Research Use Only | |
LRP2 | |
Recombinant Protein | |
Yeast | |
NCTASQFKCA SGDKCIGVTN RCDGVFDCSD NSDEAGCPTR PPGMCHSDEF QCQEDGICIP NFWECDGHPD CLYGSDEHNA CVPKTCPSSY FHCDNGNCIH RAWLCDRDND CGDMSDEKDC PTQPFRCPSW QWQCLGHNIC VNLSVVCDGI FDCPNGTDES PLCNGNSCSD FNGGCTHECV QEPFGAKCLC PLGFLLANDS KTCE | |
Greater than 90% as determined by SDS-PAGE. |