missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human L-lactate dehydrogenase C chain Protein

A DNA sequence encoding the Homo sapiens (Human) L-lactate dehydrogenase C chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6291-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16022230

  • 707.00 EUR / 100µg
Estimated Shipment: 26-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002292.1
L-lactate dehydrogenase C chain Protein
Research Use Only
LDHC
Recombinant Protein
E. coli
STVKEQLIEK LIEDDENSQC KITIVGTGAV GMACAISILL KDLADELALV DVALDKLKGE MMDLQHGSLF FSTSKITSGK DYSVSANSRI VIVTAGARQQ EGETRLALVQ RNVAIMKSII PAIVHYSPDC KILVVSNPVD ILTYIVWKIS GLPVTRVIGS GCNLDSARFR YLIGEKLGVH PTSCHGWIIG EHGDSSVPLW SGVNVAGVAL KTLDPKLGTD SDKEHWKNIH KQVIQSAYEI IKLKGYTSWA IGLSVMDLVG SILKNLRRVH PVSTMVKGLY GIKEELFLSI PCVLGRNGVS DVVKINLNSE EEALFKKSAE TLWNIQKDLI F
Greater than 90% as determined by SDS-PAGE.
3948
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers