missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLK5 Protein

A cDNA sequence encoding the KLK5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12507-HIS-INSECT-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960649

  • 186.00 EUR / 10µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

25818
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
MGSSHHHHHH SSGLVPRGSH MGSHMIINGS DCDMHTQPWQ AALLLRPNQL YCGAVLVHPQ WLLTAAHCRK KVFRVRLGHY SLSPVYESGQ QMFQGVKSIP HPGYSHPGHS NDLMLIKLNR RIRPTKDVRP INVSSHCPSA GTKCLVSGWG TTKSPQVHFP KVLQCLNISV LSQKRCEDAY PRQIDDTMFC AGDKAGRDSC QGDSGGPVVC NGSLQGLVSW GDYPCARPNR PGVYTNLCKF TKWIQETIQA NS
Greater than 90.0% as determined by SDS-PAGE.
KLK5 Protein
Research Use Only
KLK5
Human
His
The KLK5 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers