missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLK5 Protein
A cDNA sequence encoding the KLK5 was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 186.00 EUR
Specifications
Gene ID (Entrez) | 25818 |
---|---|
Name | KLK5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | KLK5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970649
|
enQuireBio™
QP12507-HIS-INSECT-2UG |
2 μg |
120.00 EUR
2µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960649
|
enQuireBio™
QP12507-HIS-INSECT-10UG |
10 μg |
186.00 EUR
10µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
25818 | |
Research Use Only | |
KLK5 | |
Human | |
His | |
The KLK5 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2M Urea and 20% glycerol. |
KLK5 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSHMIINGS DCDMHTQPWQ AALLLRPNQL YCGAVLVHPQ WLLTAAHCRK KVFRVRLGHY SLSPVYESGQ QMFQGVKSIP HPGYSHPGHS NDLMLIKLNR RIRPTKDVRP INVSSHCPSA GTKCLVSGWG TTKSPQVHFP KVLQCLNISV LSQKRCEDAY PRQIDDTMFC AGDKAGRDSC QGDSGGPVVC NGSLQGLVSW GDYPCARPNR PGVYTNLCKF TKWIQETIQA NS | |
Greater than 90.0% as determined by SDS-PAGE. |