missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL7 / Interleukin 7 Protein

Product Code. 15928418
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 15928418

Brand: enQuireBio™ QP1180100 ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_000871.1
Gene ID (Entrez) 3574
Name IL7 / Interleukin 7 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IL7
Biological Activity Shows bioactivity at a concentration of 1 ng/mL in the Mouse 2E8 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^6 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
Buffer 10 mM Aceitic acid
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.