missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-17A (Human Cell-expressed) Protein

Product Code. 18777286
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
This item is not returnable. View return policy

Product Code. 18777286

Brand: R&D Systems™ 7955IL010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Earn double rewards for reviews

  • Measured by its ability to induce CXCL1/GRO alpha secretion in HT-29 human colon adenocarcinoma cells.
  • The ED50 for this effect is 0.12 to 1.2 ng/mL.
  • Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16552
For Use With (Application) Bioactivity
Gene ID (Entrez) 3605
Molecular Weight (g/mol) 15.1 kDa (monomer)
Name CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Quantity 10 μg
Source Human embryonic kidney cell, HEK293-derived human IL-17/IL-17A protein Gly24-Ala155
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol IL17A
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.