missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HMGB1 / HMG1 Protein

A DNA sequence encoding the Homo sapiens (Human) HMGB1 / HMG1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9179-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017781

  • 560.00 EUR / 100µg
Estimated Shipment: 30-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001300821.1
HMGB1 / HMG1 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3146
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Hmgb1
Recombinant Protein
Yeast
GKGDPKKPRG KMSSYAFFVQ TCREEHKKKH PDASVNFSEF SKKCSERWKT MSAKEKGKFE DMAKADKARY EREMKTYIPP KGETKKKFKD PNAPKRPPSA FFLFCSEYRP KIKGEHPGLS IGDVAKKLGE MWNNTAADDK QPYEKKAAKL KEKYEKDIAA YRAKGKPDAA KKGVVKAEKS KKKKEEEEDE EDEEDEEEEE DEEDEDEEED DDDE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers