missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGB1 / HMG1 Protein
A DNA sequence encoding the Homo sapiens (Human) HMGB1 / HMG1, was expressed in the hosts and tags indicated.
202.00 EUR - 2155.00 EUR
Specifications
Accession Number | NP_001300821.1 |
---|---|
Gene ID (Entrez) | 3146 |
Name | HMGB1 / HMG1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017781
|
enQuireBio™
QP9179-YE-100UG |
100 μg |
565.00 EUR
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057781
|
enQuireBio™
QP9179-YE-500UG |
500 μg |
1525.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16027781
|
enQuireBio™
QP9179-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047781
|
enQuireBio™
QP9179-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067781
|
enQuireBio™
QP9179-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037781
|
enQuireBio™
QP9179-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001300821.1 | |
HMGB1 / HMG1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3146 | |
Research Use Only | |
Hmgb1 | |
Recombinant Protein | |
Yeast | |
GKGDPKKPRG KMSSYAFFVQ TCREEHKKKH PDASVNFSEF SKKCSERWKT MSAKEKGKFE DMAKADKARY EREMKTYIPP KGETKKKFKD PNAPKRPPSA FFLFCSEYRP KIKGEHPGLS IGDVAKKLGE MWNNTAADDK QPYEKKAAKL KEKYEKDIAA YRAKGKPDAA KKGVVKAEKS KKKKEEEEDE EDEEDEEEEE DEEDEDEEED DDDE | |
Greater than 90% as determined by SDS-PAGE. |